Polyethylene glycol, M.W. ≈ M.W. 600,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Granular powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %