
N-Carbethoxy-4-piperidone 98.5+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29976-53-2
:
Formula:C₈H₁₃NO₃
MDL Number:MFCD00006188
:171.19 g/mol
Boiling Pt:95…98 °C (1 mmHg)
Density:1.135 g/cm³
Flash Pt:87 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4740 to 1.4760 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- Density:(20°C) 1.13 to 1.14 kg/l
- GC:≥98.50 %