
2,6-Pyridinedicarbonyl dichloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3739-94-4
:
Formula:C₇H₃Cl₂NO₂
MDL Number:MFCD00006289
:204.006 g/mol
Boiling Pt:284 °C (760 mmHg)
Melting Pt:57…61 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:57 to 61 °C
- Titration RCOCl:≥96.0 %