Order Entry
Export
ContactUsLinkComponent
Tris(2-butoxyethyl)phosphate 95%
Tris(2-butoxyethyl)phosphate 95%
Certificates
undefined
Tris(2-butoxyethyl)phosphate 95%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4360 to 1.4390 (20°C, 589 nm)
  • GC:
    ≥94 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information