Dehydroepiandrosterone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:53-43-0
:
Formula:C₁₉H₂₈O₂
MDL Number:MFCD00003613
:288.431 g/mol
Melting Pt:146…151 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:146 to 151 °C
- Loss on drying:≤0.5 % (105 °C, 3 hrs)
- HPLC:≥98.5 %
- Specific optical rotation:+11.0° to +13.0° (20 °C, 589 nm) (c=2, Ethanol 96%)