


Tetraethylenepentamine, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-57-2
:
Formula:C₈H₂₃N₅
MDL Number:MFCD00008168
:189.31 g/mol
Boiling Pt:340 °C (760 mmHg)
Melting Pt:–40 °C
Density:0.99 g/cm³
Flash Pt:139 °C
UN:2320
ADR:8,III
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.5 %
- Titration with HCl:≥95.0 %