1,2-Diformylhydrazine 97%
Catalog # ACRO114640250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:155 to 160 °C
- TLC:≥96.0 %
Specification Test Results
About this item
:
:88.066 g/mol
Melting Pt:155…160 °C
Storage Temperature:Ambient
CAS Number:628-36-4
Formula:C₂H₄N₂O₂
MDL Number:MFCD00003275