Methyl nicotinoylacetate 95%
Catalog # ACRO425490025
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 g
- Seq:3122
Specifications
Specification Test Results
- Appearance:White to yellow to brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:34 to 42 °C
- HPLC:≥94 %
Specification Test Results
About this item
:
:179.175 g/mol
Melting Pt:34…42 °C
Storage Temperature:Ambient
CAS Number:54950-20-8
Formula:C₉H₉NO₃
MDL Number:MFCD00216524