Order Entry
Export
ContactUsLinkComponent
 
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate 98+%
 
undefined
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate 98+%

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4410 to 1.4430 (20°C, 589 nm)
  • GC:
    ≥98.0 % (sum two isomers)
  • Color scale:
    ≤10 APHA