D-γ-Tocopherol 95%
ACRO421041000
:
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
- Appearance:Clear light brown Viscous liquid
- Infrared spectrum:Conforms
- HPLC:≥94 %
:
:15,9655
:200 °C (.1 mmHg)
:>93 °C
:416.69 g/mol
:Refrigerator
:200-201-5
:54-28-4
:C₂₈H₄₈O₂
:MFCD00066529