Order Entry
Export
ContactUsLinkComponent
 
3,6-Thioxanthenediamine-10,10-dioxide 97%
 
undefined
3,6-Thioxanthenediamine-10,10-dioxide 97%

 

  • Appearance:
    Beige to yellow-green Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    214 to 219 °C
  • HPLC:
    ≥96.0 %