Order Entry
Export
Orders LinkContactUsLinkComponent
 

Pectin pract.

Pectin pract.
Certificates
Pectin pract.

3 Options Available Below

Product Details & Documents

CAS No.: 9000-69-5

  • CAS Number:9000-69-5
  • MDL Number:MFCD00081838
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    Off-white to beige to golden Powder
  • Assay:
    67 to 71 % (degree of esterification)
  • Loss on drying:
    ≤12 % (105°C, 2 hrs)
  • Particle size:
    ≤2 % ( 60 mesh)
  • Viscosity:
    30.5 to 36.5
  • pH:
    3.0 to 3.8 (1 % solution)
Product Family Options

Product Information