
Orange II, pure, certified
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:633-96-5
:
Formula:C₁₆H₁₁N₂NaO₄S
MDL Number:MFCD00011657
:350.32 g/mol
Melting Pt:164 °C
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange to dark orange-red Powder
- Infrared spectrum:Conforms
- Dye content:≥85 %
- UV Lambda max:480 to 486 nm (c=0.01 g/l, H2O)
- Absorption Ratio P-15/P+15:0.93 to 0.98
- BSC Certification:Pass