Nigrosine, water soluble, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8005-03-6
:
MDL Number:MFCD00044681
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark red to black Crystalline powder or crystals
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Titration with HClO4:≥97.5 %