Order Entry
Export
Orders LinkContactUsLinkComponent
MBD 98%

MBD 98%

MBD 98%
Certificates
MBD 98%

2 Options Available Below

Product Details & Documents

CAS No.: 33984-50-8

Synonyms:  7-(4-Methoxybenzylamino)-4-nitrobenzoxadiazole
  • CAS Number:33984-50-8
  • Formula:C₁₄H₁₂N₄O₄
  • MDL Number:MFCD00042778
  • Molecular Weight:300.274 g/mol
  • Melting Pt:176…180 °C
  • Storage Temperature:Refrigerator

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Orange to brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    177 to 182 °C
  • HPLC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 122,00 €
    122,00 €
    1 * 1 g
       122,00 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 417,00 €
    417,00 €
    1 * 5 g
       417,00 €