
Ethylene glycol dimethacrylate (ethylene dimethacrylate) 98% stabilised
:97-90-5
:
:C₁₀H₁₄O₄
:MFCD00008590
:198.21 g/mol
:98…100 °C (5 mmHg)
:–40 °C
:1.05 g/cm³
:101 °C
:GHS07
:Refrigerator
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4530 to 1.4550 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:90 to 150 ppm MEHQ