Order Entry
Export
ContactUsLinkComponent
N,N,N',N'-Tetraacetylethylenediamine 90%
N,N,N',N'-Tetraacetylethylenediamine 90%
Certificates
undefined
N,N,N',N'-Tetraacetylethylenediamine 90%

Specification Test Results

  • Appearance:
    Off-white to beige Granular powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥88 %

Specification Test Results

Related Information