
4,6-Diaminoresorcinol dihydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16523-31-2
:
Formula:C₆H₈N₂O₂
MDL Number:MFCD00143239
:213.06 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Grey-brown Crystalline solid
- Infrared spectrum:Conforms
- Titration with NaOH:≥97.5 %