Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcimycin (A23187)
Calcimycin (A23187) free acid 98%
Calcimycin (A23187) free acid 98%
:
Thermo Fisher Scientific
Calcimycin (A23187) free acid 98%
:
Thermo Fisher Scientific
:
52665-69-7
:
:
C₂₉H₃₇N₃O₆
:
MFCD00151202
:
523.63 g/mol
:
182…193 °C
:
Freezer
:
2811
:
6.1,III
:
GHS07
Appearance:
White to off-white Solid
Infrared spectrum:
Conforms
Melting point:
182 to 193 °C
TLC:
≥97.5 %
ACRO32808