Order Entry
Export
ContactUsLinkComponent
 
 
2-Methylene-1,3-propanediol 97%
 
undefined
 
2-Methylene-1,3-propanediol 97%

 

  • Appearance:
    Clear colorless to light yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4730 to 1.4750 (20°C, 589 nm)
  • GC:
    ≥96.0 %