L-Prolinamide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7531-52-4
:
Formula:C₅H₁₀N₂O
MDL Number:MFCD00005253
:114.15 g/mol
Melting Pt:97…102 °C
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:96 to 104 °C
- HPLC:≥97.5 %
- Specific optical rotation:-103° to -111° (20 °C, 589 nm) (c=1, EtOH)