
(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17553-86-5
:
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00012272
:164.204 g/mol
Melting Pt:55…64 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to yellow to beige Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:55 to 64 °C
- GC:≥88 %
- Specific optical rotation:+330° to +360° (20 °C, 589 nm) (c=1, toluene)