
2,2'-Oxybis(ethylamine) 98%
:2752-17-2
:
:C₄H₁₂N₂O
:MFCD00466591
:104.15 g/mol
:69…71 °C (15 mmHg)
:0.961 g/cm³
:Ambient
:2735
:8,III
:GHS05
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4550 to 1.4580 (20°C, 589 nm)
- GC:≥97.5 %