N-(tert-Butoxycarbonyl)-D-phenylalanine 99+%
:18942-49-9
:
:C₁₄H₁₉NO₄
:MFCD00063149
:265.31 g/mol
:78…87 °C
:Refrigerator
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:85 to 88 °C
- Water:≤0.5 % (K.F.)
- HPLC:≥99.0 %
- Specific optical rotation:-24° to -26° (20 °C, 589 nm) (c=1, ethanol)
- Titration with NaOH:≥99.0 %