
N-Boc-L-serine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3262-72-4
:
Formula:C₈H₁₅NO₅
MDL Number:MFCD00037243
:205.21 g/mol
Melting Pt:46…52 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:46 to 52 °C
- Water:7.5 to 8.0 % (K.F.)
- Specific optical rotation:-7° to -8° (20 °C, 589 nm) (c=2, H2O)
- Titration with NaOH:≥97.5 %