
Patulin 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:149-29-1
:
Formula:C₇H₆O₄
MDL Number:MFCD00005858
:154.121 g/mol
Melting Pt:108…111 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Melting point:105 to 114°C
- HPLC:≥98.5 %