
Sodium fluoride 97%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-49-4
:
Formula:NaF
MDL Number:MFCD00003524
:41.99 g/mol
Boiling Pt:1700 °C (1013 hPa)
Melting Pt:993 °C
Density:2,8 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1690
ADR:6.1,III
:GHS06
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White Powder or crystals
- Titration:≥96.0 %
- Chloride (Cl):≤120 ppm
- Sulfate (SO4):≤5000 ppm
- Sodium fluosilicate:≤1 % (Na2SiF6)
- Iron (Fe):≤200 ppm
- Heavy metals (as Pb):≤100 ppm