Industrial gloves, uvex phynomic lite
: UVEX
The uvex phynomic lite is the lightest safety glove in its class. The glove liner made of polyamide/elastane features an extremly thin aqua-polymer impregnation on palm and fingertips.
- Excellent dexterity
- Excellent dry grip
- Very high level of breathability
- Coating is extremely thin but still highly durable
Ideal for precision assembly work and other precision work. Suitable for use in dry and slightly damp areas.
: EN 388:2016 (2121X)
Food contact approved
OEKO-TEX® Standard 100
Food contact approved
OEKO-TEX® Standard 100
- Catalog No:
- 112-4505
- 112-4504
- 112-4328
- 112-4327
- 112-4340
- 112-4329
- Model:
- lite
- lite
- lite
- lite
- lite
- lite
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Finish:
- Aqua-polymer impregnation on palm and fingertips
- Aqua-polymer impregnation on palm and fingertips
- Aqua-polymer impregnation on palm and fingertips
- Aqua-polymer impregnation on palm and fingertips
- Aqua-polymer impregnation on palm and fingertips
- Aqua-polymer impregnation on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- Food contact approved:
- +
- +
- +
- +
- +
- +