ContactUsLinkComponent
Cold resistant gloves, uvex unilite thermo
Cold resistant gloves, uvex unilite thermo
Supplier:  UVEX
Certificates
undefined
Cold resistant gloves, uvex unilite thermo
Supplier:  UVEX
Specifications
  • Catalog No:
  • 112-4535
  • 112-4534
  • 112-4532
  • 112-4531
  • 112-4533
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Finish:
  • Polymer coating on palm and fingertips
  • Polymer coating on palm and fingertips
  • Polymer coating on palm and fingertips
  • Polymer coating on palm and fingertips
  • Polymer coating on palm and fingertips
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3131X), EN 511 (010)
  • EN 388:2016 (3131X), EN 511 (010)
  • EN 388:2016 (3131X), EN 511 (010)
  • EN 388:2016 (3131X), EN 511 (010)
  • EN 388:2016 (3131X), EN 511 (010)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +