Orders LinkContactUsLinkComponent
Biotin NTA
Biotin NTA
 :   Biotium
Biotin NTA
 :  Biotium
Certificates
Biotin NTA
2 Options Available Below
Biotinylated probes for detection of His-tagged proteins immobilised on nitrocellulose membranes.
  • White solid soluble in water
  • Removable from the histidine-tagged protein at pH 4,8
Biotin NTA, also known as BNTA (Biotin nitrilotriacetic acid, tripotassium salt) reversibly binds to histidine-tagged proteins, and can be detected with Biotium's CF™ Dye streptavidin or monoclonal anti-biotin fluorescent conjugates. Available with 6-atom aminohexanoyl (X) linker between the biotin and NTA to facilitate interaction with streptavidin.
Product Family Options
Product Information
  • Description
    Pk
  • Biotin NTA or BNTA (Biotin nitrilotriacetic acid, tripotassium salt)
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 197,00 €
    197,00 €
    1 * 5 mg
       197,00 €
  • Biotin-X NTA (Biotin-X nitrilotriacetic acid, tripotassium salt)
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 197,00 €
    197,00 €
    1 * 5 mg
       197,00 €