Orders LinkContactUsLinkComponent
Heating tapes, IT-S45
 :   ISOPAD
Heating tapes, IT-S45
 :  ISOPAD
Heating tapes, IT-S45
Heating tapes, IT-S45
Isopad IT-S45 is a preterminated, flexible heating tape consisting of glassfibre insulated, flexible fabric.
  • Maximum operating temperature: 450 °C
  • Width×Thickness: 30×5 mm (±10%)
  • Maximum power per meter 250 W/m
The protective overbraid of the coiled heating conductors meets the requirements of Protection Class I. This heating tape is typically used for heating up and compensating for heat losses on short pipes and small sized structures. The heating tape is not moisture protected and must only be used in dry areas.
Product Family Options
  • Description
  • Heating tape IT-S45, 0,5 m length, 100 W
    1 * 1 items
    1 * 1 items - 198,20 €
    198,20 €
    1 * 1 items
       198,20 €
  • Heating tape IT-S45, 1 m length, 250 W
    1 * 1 items
    1 * 1 items - 210,20 €
    210,20 €
    1 * 1 items
       210,20 €
  • Heating tape IT-S45, 1,5 m length, 375 W
    1 * 1 items
    1 * 1 items - 239,60 €
    239,60 €
    1 * 1 items
       239,60 €
  • Heating tape IT-S45, 2 m length, 500 W
    1 * 1 items
    1 * 1 items - 276,60 €
    276,60 €
    1 * 1 items
       276,60 €
  • Heating tape IT-S45, 3 m length, 750 W
    1 * 1 items
    1 * 1 items - 341,00 €
    341,00 €
    1 * 1 items
       341,00 €
  • Heating tape IT-S45, 5 m length, 1250 W
    1 * 1 items
    1 * 1 items - 463,50 €
    463,50 €
    1 * 1 items
       463,50 €