ContactUsLinkComponent
Cryogenic vial grippers, Corning®
Cryogenic vial grippers, Corning®
  479-1846
 :  Corning
undefined
Cryogenic vial grippers, Corning®
  479-1846
 :  Corning
  • Description:
    Cryogenic vial grippers, assorted colours
  • Seq:
    01
  • Pk:
    5
These cryogenic vial grippers feature a unique design to grasp internal or external-thread cryogenic vials.
They enable to easily sort or move vials while maintaining sterility and protecting fingers from frozen vials, dry ice and liquid nitrogen.