Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dexamethasone sodium phosphate
Dexamethasone sodium phosphate ≥97%
Dexamethasone sodium phosphate ≥97%
Dexamethasone sodium phosphate ≥97%
Supplier:
MP Biomedicals
Certificates
Expand image
Dexamethasone sodium phosphate ≥97%
Supplier:
MP Biomedicals
About this item
CAS Number:
2392-39-4
:
Formula:
C₂₂H₂₈FNa₂O₈P
MDL Number:
MFCD00079105
:
516.41 g/mol