Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
γ-L-Glutamyl-p-nitroanilide hydrochloride
γ-L-Glutamyl-p-nitroanilide hydrochloride, beige powder
γ-L-Glutamyl-p-nitroanilide hydrochloride, beige powder
:
MP Biomedicals
γ-L-Glutamyl-p-nitroanilide hydrochloride, beige powder
:
MP Biomedicals
:
67953-08-6
:
:
C₁₁H₁₄ClN₃O₅
:
MFCD00039054
:
303.699 g/mol
Identity Test:
Passes
Optical Rotation (c=1, 0.1 N HCl):
+30° ± 5°
UV/Vis: λmax (water):
317 ± 5 nm