Pectin, powder
Supplier: MP Biomedicals
Jump to:
About this item
Soluble in water (1 g/20 ml - viscous solution acid to litmus); insoluble in organic solvents.
Pectin is a heterosaccharide, component of terrestrial plant cell walls.
It is used as a substrate to identify, differentiate and characterised pectinase(s). It is also used to study its degradation by pectinolytic bacteria.
- CAS Number:9000-69-5
- MDL Number:MFCD00081838
3 Options Available Below
Product Details & Documents
Soluble in water (1 g/20 ml - viscous solution acid to litmus); insoluble in organic solvents.
Pectin is a heterosaccharide, component of terrestrial plant cell walls.
It is used as a substrate to identify, differentiate and characterised pectinase(s). It is also used to study its degradation by pectinolytic bacteria.
- CAS Number:9000-69-5
- MDL Number:MFCD00081838



