Ammonia TNT plus kit
Catalog # HACHTNT830
Supplier: Hach
CAS Number:
Specifications
- Pk:25
- Description:Ammonia TNT plus kit, ULR
- Seq:01
- Range:0,015 - 2 mg/l
Specifications
About this item
This kit is used for the determination of ammonia nitrogen by the salicylate method using TNTplus vials.
- Reduce errors with bar coded vials and RFID
- Innovative Vial
- No reagent blank necessary