Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Valine USP

L(+)-Valine USP

L(+)-Valine USP
Supplier:  Spectrum Chemical
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  H-Val-OH, Valine, (S)-(+)-Valine
  • CAS Number:72-18-4
  • Formula:C₅H₁₁NO₂
  • MDL Number:MFCD00064220
  • Molecular Weight:117.15 g/mol
  • Merck Index:13,09458
  • Boiling Pt:217 °C (1013 hPa)
  • Melting Pt:86 °C
  • Density:1.284 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-773-6

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    98.5% Min.
Product Family Options

Product Information