D-(+)-Biotin, powder USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:58-85-5
:
Formula:C₁₀H₁₆N₂O₃S
MDL Number:MFCD00005541
:244.31 g/mol
Merck Index:13,01231
Melting Pt:232…233 °C
Density:1.41 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:200-399-3
Specification Test Results
- Assay:97.5% Min.