L(+)-Serine USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:56-45-1
:
Formula:C₃H₇NO₃
MDL Number:MFCD00064224
:105.09 g/mol
Merck Index:14,00846
Boiling Pt:259 °C (1013 hPa)
Melting Pt:215…225 °C
Density:1.6 g/cm³ (22 °C)
Storage Temperature:Ambient
EINECS:200-274-3
Specification Test Results
- Assay:98.5% Min.