Order Entry
Export
Orders LinkContactUsLinkComponent
Dextrin, yellow, powder

Dextrin, yellow, powder

Supplier:   Spectrum Chemical
Dextrin, yellow, powder
Supplier:  Spectrum Chemical
Certificates
Dextrin, yellow, powder

2 Options Available Below

Product Details & Documents

Form: Yellow Powder

Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Storage Temperature:Ambient
  • EINECS:232-675-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 371,00 €
    371,00 €
    1 * 2,5 kg
       371,00 €

  • 50 lb
    Log in for availability
    Log in for availability
    1 * 50 lb
    1 * 50 lb - 1 180,00 €
    1 180,00 €
    1 * 50 lb
       1 180,00 €