Butyl isocyanate ≥97%
Catalog # L05664.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:99.13 g/mol
Merck Index:14,1575
Boiling Pt:112…116 °C
Melting Pt:–75 °C
Density:0.890
Flash Pt:11 °C (51 °F)
Storage Temperature:Refrigerator
EINECS:203-862-8
UN:2485
ADR:6.1,I
:GHS02
:GHS06
:GHS08
:GHS05
CAS Number:111-36-4
Formula:C₅H₉NO
MDL Number:MFCD00002046