
2,4-Difluorophenylglyoxal hydrate 95% (dry weight)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79784-36-4
:
Formula:C₈H₆F₂O₃
MDL Number:MFCD04038287
:188.13 g/mol
Melting Pt:51…54 °C
:GHS07
Storage Temperature:Ambient