Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-2,2-diethylbutyrate
Ethyl-2,2-diethylbutyrate 98%
Ethyl-2,2-diethylbutyrate 98%
L19972.06
:
Thermo Fisher Scientific
:
Ethyl-2,2-diethylbutyrate 98%
L19972.06
:
Thermo Fisher Scientific
:
Pk:
5 g
Seq:
3173
:
:
172.27 g/mol
:
85…87 °C (30 mmHg)
:
0.884
:
Ambient
:
34666-17-6
:
C₁₀H₂₀O₂
:
MFCD00154424