
N-Boc-1,6-diaminohexane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51857-17-1
:
Formula:C₁₁H₂₄N₂O₂
MDL Number:MFCD00671489
:216.33 g/mol
Boiling Pt:106…110 °C (0.3 mmHg)
Density:0.965
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
ADR:8,III
:GHS05