Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,N'-Dibenzylethylenediamine diacetate
N,N'-Dibenzylethylenediamine diacetate 99%
N,N'-Dibenzylethylenediamine diacetate 99%
:
Thermo Fisher Scientific
N,N'-Dibenzylethylenediamine diacetate 99%
:
Thermo Fisher Scientific
:
122-75-8
:
:
C₁₆H₂₀N₂
:
MFCD00040588
:
14,1064
:
117…118 °C
:
360.45 g/mol
:
204-572-4
:
GHS06
:
Ambient
msds_ALFAL17974
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years