Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dowex® 50WX2 (H)
Dowex® 50WX2 (H) 50-100 mesh
Dowex® 50WX2 (H) 50-100 mesh
Dowex® 50WX2 (H) 50-100 mesh
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Dowex® 50WX2 (H) 50-100 mesh
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
12612-37-2
:
69011-20-7
MDL Number:
MFCD00132722
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL13943
Videos
Thermo Scientific Acroseal 25 Years
Documentation