
1,4-Bis(3-aminopropyl)piperazine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7209-38-3
:
Formula:C₁₀H₂₄N₄
MDL Number:MFCD00006161
:200.33 g/mol
Boiling Pt:150…152 °C (2 mmHg)
Melting Pt:14 °C
Density:0.973
Flash Pt:162 °C (323 °F)
Storage Temperature:Ambient
EINECS:230-589-1
UN:2735
ADR:8,II
:GHS05