


4,4'-Methylenebis(cyclohexylamine) (mixture of isomers) mixture of stereoisomers ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1761-71-3
:
Formula:C₁₃H₂₆N₂
MDL Number:MFCD00001496
Boiling Pt:325…327 °C
Melting Pt:∼ 43 °C
Flash Pt:159 °C (318 °F)
Storage Temperature:Ambient
EINECS:217-168-8
UN:2928
ADR:6.1,II
:210.37 g/mol
:GHS06
:GHS05
:GHS07