
2,2-Di-n-propylacetamide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2430-27-5
:
Formula:C₈H₁₇NO
MDL Number:MFCD00051534
:143.23 g/mol
Merck Index:14,9914
Melting Pt:123…127 °C
EINECS:219-394-2
:GHS07
Storage Temperature:Ambient