4,6-Diamino-2-(methylthio)pyrimidine 97%
Catalog # L07888.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:1005-39-6
:
Formula:C₅H₈N₄S
MDL Number:MFCD00023241
:156.21 g/mol
Melting Pt:187…191 °C
Storage Temperature:Ambient
EINECS:213-735-9